Please login to your online account to display your discounted pricing

anti-MICB, Alexa Fluor 488, Clone: 236511, R&D Systems


Manufacturer: r & d systems  RDSFAB1599G025

 View more versions of this product

Catalog No. FAB1599G025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen MICB
Quantity 25 Tests
Cross Reactivity Human
Format Purified
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MICB. Ala23-Gly298
Isotype IgG2b
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 236511
Conjugate Alexa Fluor 488
Purification Method Protein A or G purified from hybridoma culture supernatant