Please login to your online account to display your discounted pricing

anti-MICA, PE, Clone: 159227, R&D Systems


Manufacturer: r & d systems  RDSFAB1300P100

 View more versions of this product

Catalog No. FAB1300P100

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2b
Regulatory Status RUO
Format Purified
Cross Reactivity Human
Quantity 100 Tests
Clone 159227
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PE
Immunogen BaF3 mouse pro-B cell line transfected with human MICA.
Host Species Mouse
Antigen MICA
Purification Method Protein A or G purified from hybridoma culture supernatant