Please login to your online account to display your discounted pricing

anti-MICA, PE, Clone: 159227, R&D Systems


Manufacturer: r & d systems  RDSFAB1300P025

 View more versions of this product

Catalog No. FAB1300P025

  • / Each
 

Description & Specifications

Specifications

Antigen MICA
Clone 159227
Conjugate PE
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen BaF3 mouse pro-B cell line transfected with human MICA.
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human