Please login to your online account to display your discounted pricing

anti-MICA, APC, Clone: 159227, R&D Systems


Manufacturer: r & d systems  RDSFAB1300A100

 View more versions of this product

Catalog No. FAB1300A100

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2b
Format Purified
Host Species Mouse
Cross Reactivity Human
Immunogen BaF3 mouse pro-B cell line transfected with human MICA.
Antigen MICA
Primary or Secondary Primary
Conjugate APC
Monoclonal or Polyclonal Monoclonal
Quantity 100 Tests
Regulatory Status RUO
Clone 159227
Purification Method Protein A or G purified from hybridoma culture supernatant