Please login to your online account to display your discounted pricing

anti-MICA, Alexa Fluor 488, Clone: 159227, R&D Systems


Manufacturer: r & d systems  RDSFAB1300G100

 View more versions of this product

Catalog No. FAB1300G100

  • / Each
 

Description & Specifications

Specifications

Antigen MICA
Format Purified
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human MICA.
Isotype IgG2b
Regulatory Status RUO
Cross Reactivity Human
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 159227
Conjugate Alexa Fluor 488