Please login to your online account to display your discounted pricing

anti-MGLUR4 Polyclonal, Thermo Scientific™

MGLUR4 Polyclonal antibody PA5-33822 from Thermo Scientific™ specifically detects MGLUR4 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

$509.25 - $509.25

Specifications

Antigen MGLUR4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GRM4/MGLUR4.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533822

 
pierce biotechnology inc
PA533822
Unlabeled 50μg Each for $509.25

PIPA533821

 
pierce biotechnology inc
PA533821
Unlabeled 50μg Each for $509.25
Description & Specifications

Specifications

Antigen MGLUR4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human GRM4/MGLUR4.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

MGLUR4 Polyclonal antibody PA5-33822 from Thermo Scientific™ specifically detects MGLUR4 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

FSHR, a Glycoprotein Hormone Receptor, is required for normal ovarian development and follicle maturation in females. When activated, it stimulates ovarian follicular growth and production of estrogens, initiating a normal menstrual cycle. In males, FSHR is required for the initiation of spermatogenesis and normal sperm production. Two isoforms are produced by alternative splicing. FSHR expression has been reported in ovary, uterus, and testis. ESTs have been isolated from testis libraries.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4/MGLUR4.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: GRM4, GPRC1D, MGlu4, MGLUR4
Uniprot Accession: Q14833
Gene Symbol: GRM4
Entrez GeneID: 2914
PA5-33547 detects FSH Receptor/FSHR in Human samples. PA5-33547 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-33547 The immunogen is Synthetic 17 amino acid peptide from N-terminal extracellular domain of human FSHR..