Please login to your online account to display your discounted pricing

anti-MGLUR4 Polyclonal, Thermo Scientific™


MGLUR4 Polyclonal antibody PA5-33822 from Thermo Scientific™ specifically detects MGLUR4 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

Manufacturer: pierce biotechnology inc  PA533822

 View more versions of this product

Catalog No. PIPA533822

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene ID (Entrez) 2914
Applications Immunohistochemistry (Paraffin)
Gene Accession No. Q14833
Gene Alias GRM4, GPRC1D, MGlu4, MGLUR4
Gene Symbols GRM4
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4/MGLUR4.
Species Reactivity Human
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Antigen MGLUR4
Format Purified
Isotype IgG
Quantity 50μg
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal

MGLUR4 Polyclonal antibody PA5-33822 from Thermo Scientific™ specifically detects MGLUR4 in Human samples, and it is validated for Immunohistochemistry (Paraffin)

The Metabotropic Glutamate Receptor mGluR4/GRM4 binds L-glutamate, the major excitatory neurotransmitter in the central nervous system, which activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4/MGLUR4.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: GRM4, GPRC1D, MGlu4, MGLUR4
Uniprot Accession: Q14833
Entrez GeneID: 2914
Gene Symbol: GRM4
This product detects MGLUR4 in Human samples. This product has been successfully used in Immunohistochemistry (Paraffin) procedures. The immunogen is Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4/MGLUR4..