Please login to your online account to display your discounted pricing

anti-Methionine Sulfoxide, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP106707

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP106707

  • / Each
 

Description & Specifications

Specifications

Test Specificity Specific for all methionine sulfoxide containing proteins and peptides.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Antigen Methionine Sulfoxide
Regulatory Status RUO
Immunogen Oxidized methionine rich zein protein from Zea mays.
Format Antiserum
Host Species Rabbit