Please login to your online account to display your discounted pricing

anti-Methionine Sulfoxide, Polyclonal, Novus Biologicals™

$321.00 - $321.00

Specifications

Host Species Rabbit
Immunogen Oxidized methionine rich zein protein from Zea mays.
Antigen Methionine Sulfoxide
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP106707

 
novus biologicals canada
NBP106707
Unlabeled 0.1mL Each for $321.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Oxidized methionine rich zein protein from Zea mays.
Antigen Methionine Sulfoxide
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Specific for all methionine sulfoxide containing proteins and peptides.