Please login to your online account to display your discounted pricing

anti-Mer, Polyclonal, Novus Biologicals™

$139.00 - $300.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human MERTK was used as the immunogen.
Antigen Mer
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224650SS

 
novus biologicals canada
NBP224650SS
Unlabeled 0.025mg Each for $139.00

NBP224650

 
novus biologicals canada
NBP224650
Unlabeled 0.1mg Each for $300.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human MERTK was used as the immunogen.
Antigen Mer
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, dog, rat, mouse, chicken and zebrafish and 92% homologous in xenopus.
Gene Accession No. Q12866
Gene ID (Entrez) 10461
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL