Please login to your online account to display your discounted pricing

anti-Mer, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224650SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224650SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q12866
Gene ID (Entrez) 10461
Primary or Secondary Primary
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, dog, rat, mouse, chicken and zebrafish and 92% homologous in xenopus.
Host Species Rabbit
Antigen Mer
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Quantity 0.025mg
Format Purified
Immunogen A portion of amino acids 50-100 of human MERTK was used as the immunogen.
Applications Western Blotting
Regulatory Status RUO