Please login to your online account to display your discounted pricing

anti-Mer, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224650

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224650

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q12866
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10461
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, dog, rat, mouse, chicken and zebrafish and 92% homologous in xenopus.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Antigen Mer
Format Purified
Quantity 0.1mg
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human MERTK was used as the immunogen.
Regulatory Status RUO