Please login to your online account to display your discounted pricing

anti-Melatonin Receptor 1B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NLS932SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NLS932SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P49286
Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4544
Antigen Melatonin Receptor 1B
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286].
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL