missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-MEKK2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of human MEKK2 was used as the immunogen.
Antigen MEKK2
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224756 Novus Biologicals Canada
NBP224756
0.1mg Each for $255.80
NBP224756SS Novus Biologicals Canada
NBP224756SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 1-50 of human MEKK2 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
10746
0.5mg/mL
IgG
MEKK2
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (CRA_a and CRA_c) and chimpanzee, 88% homologous in hamster, mouse and rat, 82% homologous in bovine and horse, and 75% homologous in dog.
100μg in 200μL PBS containing 0.2% gelatin