Please login to your online account to display your discounted pricing

anti-MEKK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224756SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224756SS

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Primary or Secondary Primary
Gene ID (Entrez) 10746
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (CRA_a and CRA_c) and chimpanzee, 88% homologous in hamster, mouse and rat, 82% homologous in bovine and horse, and 75% homologous in dog.
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Antigen MEKK2
Immunogen A portion of amino acids 1-50 of human MEKK2 was used as the immunogen.
Host Species Rabbit
Format Purified
Cross Reactivity Human