Please login to your online account to display your discounted pricing

anti-MEKK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224756

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224756

  • / Each
 

Description & Specifications

Specifications

Antigen MEKK2
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human MEKK2 was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10746
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (CRA_a and CRA_c) and chimpanzee, 88% homologous in hamster, mouse and rat, 82% homologous in bovine and horse, and 75% homologous in dog.
Target Species Human