Please login to your online account to display your discounted pricing

anti-MEK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056449SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056449T

  • / Each
 

Description & Specifications

Specifications

Antigen MEK2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P36507
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human MEK2.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5605
Test Specificity In 293, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat based on sequence identity.
Target Species Human