Please login to your online account to display your discounted pricing

anti-MEK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056449

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056449

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P36507
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Gene ID (Entrez) 5605
Test Specificity In 293, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat based on sequence identity.
Concentration 0.5mg/mL
Conjugate Unlabeled
Applications Western Blotting
Immunogen This antibody was developed against a synthetic peptide of human MEK2.
Monoclonal or Polyclonal Polyclonal
Antigen MEK2
Format Purified
Host Species Rabbit
Cross Reactivity Human
Quantity 0.1mg
Regulatory Status RUO