Please login to your online account to display your discounted pricing

anti-MEK2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human MEK2.
Antigen MEK2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056449

 
novus biologicals canada
NB10056449
Unlabeled 0.1mg Each for $255.80

NB10056449T

 
novus biologicals canada
NB10056449SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide of human MEK2.
Antigen MEK2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In 293, a 45 kDa band is observed. Cross reacts with Human. Predicted to cross react with Mouse and Rat based on sequence identity.
Gene Accession No. P36507
Gene ID (Entrez) 5605
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL