Please login to your online account to display your discounted pricing

anti-MEK1, Polyclonal, Novus Biologicals™

$139.00 - $332.66

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250
Antigen MEK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224542

 
novus biologicals canada
NBP224542
Unlabeled 0.05mg Each for $332.66

NBP224542SS

 
novus biologicals canada
NBP224542SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 200-250
Antigen MEK1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this RXFP1 antibody is 100% homologous in human, chimp and orangutan, and 83% homologous in horse, bovine, dog and cat.
Gene ID (Entrez) 5604
Formulation 50μg phospho antibody in 100μL PBS containing 0.05% BSA
Concentration 0.5mg/mL