Please login to your online account to display your discounted pricing

anti-MEIS1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 280-330 of MEIS1 was used as the immunogen.
Antigen MEIS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224522

 
novus biologicals canada
NBP224522
Unlabeled 0.1mg Each for $255.80

NBP224522SS

 
novus biologicals canada
NBP224522SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 280-330 of MEIS1 was used as the immunogen.
Antigen MEIS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat, mouse, monkey, cow, and dog, and 93% homologous in horse.
Gene Accession No. O00470
Gene ID (Entrez) 4211
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL