Please login to your online account to display your discounted pricing

anti-MEIS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224522

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224522

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4211
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat, mouse, monkey, cow, and dog, and 93% homologous in horse.
Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Conjugate Unlabeled
Concentration 0.5mg/mL
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Xenopus
Gene Accession No. O00470
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mg
Host Species Rabbit
Format Affinity Purified
Immunogen A portion of amino acids 280-330 of MEIS1 was used as the immunogen.
Regulatory Status RUO
Antigen MEIS1