Please login to your online account to display your discounted pricing

anti-MDM2/HDM2, Polyclonal, Novus Biologicals™

$109.00 - $300.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MDM2 (within residues 300-400). [Swiss-Prot No. Q00987]
Antigen MDM2/HDM2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP102158

 
novus biologicals canada
NBP102158
Unlabeled 0.1mL Each for $300.00

NBP102158SS

 
novus biologicals canada
NBP102158SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MDM2 (within residues 300-400). [Swiss-Prot No. Q00987]
Antigen MDM2/HDM2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Human. Predicted to react with monkey based on 100% sequence identity.
Gene Accession No. Q00987
Gene ID (Entrez) 4193
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL