Please login to your online account to display your discounted pricing

anti-MDM2/HDM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP102158SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP102158SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q00987
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity Human. Predicted to react with monkey based on 100% sequence identity.
Gene ID (Entrez) 4193
Concentration 1mg/mL
Conjugate Unlabeled
Immunogen Synthetic peptide made to an internal portion of human MDM2 (within residues 300-400). [Swiss-Prot No. Q00987]
Cross Reactivity Human
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mL
Regulatory Status RUO
Antigen MDM2/HDM2
Format Affinity Purified