Please login to your online account to display your discounted pricing

anti-MDM2/HDM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP102158

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP102158

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4193
Gene Accession No. Q00987
Primary or Secondary Primary
Test Specificity Human. Predicted to react with monkey based on 100% sequence identity.
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Antigen MDM2/HDM2
Formulation Tris-citrate/phosphate, pH 7-8
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of human MDM2 (within residues 300-400). [Swiss-Prot No. Q00987]
Quantity 0.1mL
Regulatory Status RUO
Cross Reactivity Human