Please login to your online account to display your discounted pricing

anti-MDC1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 871-888 (RQESDKNGESASPERDRE) of human NFBD1.
Antigen MDC1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056657

 
novus biologicals canada
NB10056657
Unlabeled 0.2mL Each for $255.80

NB10056657T

 
novus biologicals canada
NB10056657SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 871-888 (RQESDKNGESASPERDRE) of human NFBD1.
Antigen MDC1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In Jurkat, an approx. 220 kDa band is observed.
Gene Accession No. NP_055456
Gene ID (Entrez) 9656
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL