Please login to your online account to display your discounted pricing

anti-MDC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056657

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056657

  • / Each
 

Description & Specifications

Specifications

Antigen MDC1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_055456
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 871-888 (RQESDKNGESASPERDRE) of human NFBD1.
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9656
Test Specificity In Jurkat, an approx. 220 kDa band is observed.
Target Species Human