Please login to your online account to display your discounted pricing

anti-MD-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056655SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056655T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. NP_056179
Gene ID (Entrez) 23643
Concentration 0.5mg/mL
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Format Purified
Host Species Rabbit
Antigen MD-2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Immunogen A full length recombinant protein was used as the immunogen.