Please login to your online account to display your discounted pricing

anti-MD-1/LY86, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056700

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056700

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity In Ramos, a 25 kDa band is observed.
Gene Accession No. O95711
Primary or Secondary Primary
Concentration 0.5mg/mL
Gene ID (Entrez) 9450
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Host Species Rabbit
Quantity 0.1mg
Antigen MD-1/LY86
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was developed against a KLH-conjugated synthetic peptide corresponding to amino acids 112-125 of human MD-1.
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO