Please login to your online account to display your discounted pricing

anti-MCT1/SLC16A1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP159656

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP159656

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 1.0mg/mL
Formulation PBS
Primary or Secondary Primary
Gene ID (Entrez) 6566
Conjugate Unlabeled
Format Affinity Purified
Gene Accession No. P53985
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A synthetic peptide made to an internal portion of the human Monocarboxylic acid transporter 1 protein (between residues 200-300) [UniProt P53985]
Host Species Rabbit
Quantity 0.1mg
Antigen MCT1/SLC16A1
Regulatory Status RUO