Please login to your online account to display your discounted pricing

anti-mCherry, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225158SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225158SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity This antibody detects mCherry.
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity All
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Precipitation
Immunogen Recombinant full length His tagged mCherry purified from E. coli
Quantity 0.025mL
Host Species Chicken
Antigen mCherry