missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-MCAM/CD146, Fluorescein, Clone: 128018, R&D Systems

$353.45 - $353.45

Specifications

Clone 128018
Host Species Murine
Isotype IgG1
Immunogen Mouse myeloma cell line NS0-derived recombinant human MCAM/CD146
Antigen MCAM/CD146
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FAB932F100 R & D Systems
FAB932F100
100 Tests Each for $353.45
Specifications Description & Specifications

Specifications

128018
IgG1
MCAM/CD146
Purified
RUO
Detects human MCAM/CD146 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm)OCAM, or rmMAdCAM-1.
4162
Protein A or G purified from hybridoma culture supernatant
Murine
Mouse myeloma cell line NS0-derived recombinant human MCAM/CD146
Monoclonal
Primary
Flow Cytometry
AAA20824
Supplied in a saline solution containing BSA and Sodium Azide.