Please login to your online account to display your discounted pricing

anti-MCAM/CD146, Fluorescein, Clone: 128018, R&D Systems


Manufacturer: r & d systems  FAB932F100

 View more versions of this product

Catalog No. FAB932F100

  • / Each
 

Description & Specifications

Specifications

Immunogen Mouse myeloma cell line NS0-derived recombinant human MCAM/CD146
Antigen MCAM/CD146
Format Purified
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. AAA20824
Host Species Mouse
Applications Flow Cytometry
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4162
Test Specificity Detects human MCAM/CD146 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm)OCAM, or rmMAdCAM-1.
Clone 128018
Conjugate Fluorescein
Cross Reactivity Human