Please login to your online account to display your discounted pricing

anti-MBD4, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 268-300 and 337-374 of human MBD4.
Antigen MBD4
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224393SS

 
novus biologicals canada
NBP224393SS
Unlabeled 0.025mg Each for $109.00

NBP224393

 
novus biologicals canada
NBP224393
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 268-300 and 337-374 of human MBD4.
Antigen MBD4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene ID (Entrez) 8930
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL