Please login to your online account to display your discounted pricing

anti-MBD4, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243]
Antigen MBD4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP191189

 
novus biologicals canada
NBP191189
Unlabeled 0.1mL Each for $309.05

NBP191189SS

 
novus biologicals canada
NBP191189SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243]
Antigen MBD4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human. Does not react with mouse.
Gene Accession No. O95243
Gene ID (Entrez) 8930
Formulation PBS, 30% glycerol
Concentration 1.11mg/mL