Please login to your online account to display your discounted pricing

anti-MAT2A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11094158

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11094158

  • / Each
 

Description & Specifications

Specifications

Antigen MAT2A
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Concentration 1.64mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P31153
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). [Swiss-Prot No. P31153]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4144
Test Specificity Human, rat, mouse, orangutan, bovine and monkey. Immunogen sequence has 94% homology to porcine and chicken and 86% homology to Zebrafish.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat