Please login to your online account to display your discounted pricing

anti-MAT1/2A, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot No. P31153]
Antigen MAT1/2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11094162T

 
novus biologicals canada
NB11094162SS
Unlabeled 0.025mL Each for $109.00

NB11094162

 
novus biologicals canada
NB11094162
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot No. P31153]
Antigen MAT1/2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Test Specificity Human, rat, orangutan, bovine, Zebrafish and monkey. Does not react with mouse.
Gene Accession No. P31153
Gene ID (Entrez) 4144
Formulation PBS, 30% glycerol
Concentration 1mg/mL