Please login to your online account to display your discounted pricing

anti-Mas, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NLS1531

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NLS1531

  • / Each
 

Description & Specifications

Specifications

Antigen Mas
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, (pH7.7)
Gene Accession No. P04201
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide made to the C-terminus of human MAS1. [UniProt No. P04201]
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4142
Test Specificity Human and mouse. Immunogen sequence has 82% identity with rat.
Target Species Human
Target Species Murine