missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-MAPRE1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 228-261 of human MAPRE1. Genbank accession no. NM_012325.
Antigen MAPRE1
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224946 Novus Biologicals Canada
NBP224946
0.1mg Each for $257.41
NBP224946SS Novus Biologicals Canada
NBP224946SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
A synthetic peptide corresponding to amino acids 228-261 of human MAPRE1. Genbank accession no. NM_012325.
Polyclonal
Primary
Western Blot
22919
IgG
MAPRE1
Purified
RUO
Q15691
100μg in 200mL PBS containing 0.2% gelatin