Please login to your online account to display your discounted pricing

anti-MAPRE1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224946

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224946

  • / Each
 

Description & Specifications

Specifications

Antigen MAPRE1
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200mL PBS containing 0.2% gelatin
Gene Accession No. Q15691
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 228-261 of human MAPRE1. Genbank accession no. NM_012325.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 22919
Target Species Human