Please login to your online account to display your discounted pricing

Map1lc3b Polyclonal Antibody, Invitrogen™


Map1lc3b Antibody, OSM00026W, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSM00026W

Catalog No. PIOSM00026W

  • / Each
 

Description & Specifications

Specifications

Antigen Map1lc3b
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Serum
Formulation 150μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q9GZQ8
Gene Alias MAP1A/MAP1B LC3 B, Map1lc3b
Gene Symbols MAP1LC3B
Host Species Rabbit
Immunogen A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3B
Quantity 150μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 81631
Target Species Human

Map1lc3b Antibody, OSM00026W, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3B
Formulation: 150μL unpurified antibody in whole serum and no preservative
Synonyms: MAP1A/MAP1B LC3 B, Map1lc3b
Uniprot Accession: Q9GZQ8
Entrez GeneID: 81631
Gene Symbol: MAP1LC3B
Reconstitute with 150 ul of distilled water.