Please login to your online account to display your discounted pricing

anti-Map1lc3b Polyclonal, Thermo Scientific™

Map1lc3b Polyclonal antibody OSM00026W from Thermo Scientific™ specifically detects Map1lc3b in Human samples, and it is validated for Immunohistochemistry and Western Blot

$448.05 - $448.05

Specifications

Antigen Map1lc3b
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSM00025W

 
pierce biotechnology inc
OSM00025W
Unlabeled 150μL Each for $448.05

PIOSM00026W

 
pierce biotechnology inc
OSM00026W
Unlabeled 150μL Each for $448.05

PIOSM00027W

 
pierce biotechnology inc
OSM00027W
Unlabeled 150μL Each for $448.05
Description & Specifications

Specifications

Antigen Map1lc3b
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from mouse microtubule-associated proteins 1A/1B light chain 3B
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Map1lc3b Polyclonal antibody OSM00026W from Thermo Scientific™ specifically detects Map1lc3b in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3B
Formulation: 150μL unpurified Antibody in whole serum, with no preservative;
Synonyms: MAP1A/MAP1B LC3 B, Map1lc3b
Uniprot Accession: Q9GZQ8
Gene Symbol: MAP1LC3B
Entrez GeneID: 81631
OSM00026W detects Map1lc3b in Human samples. OSM00026W has been successfully used in Immunohistochemistry and Western Blot procedures. OSM00026W The immunogen is A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3B.