Please login to your online account to display your discounted pricing

Map1lc3b Polyclonal Antibody, Invitrogen™


Map1lc3b Antibody, OSM00025W, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSM00025W

Catalog No. PIOSM00025W

  • / Each
 

Description & Specifications

Specifications

Formulation 150μL unpurified antibody in whole serum and no preservative
Gene Symbols MAP1LC3B
Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Mouse
Cross Reactivity Rat
Storage Requirements avoiding freeze/thaw cycles.
Gene ID (Entrez) 64862, 67443
Format Serum
Gene Alias MAP1A/MAP1B LC3 B, Map1lc3b
Gene Accession No. Q62625, Q9CQV6
Species Reactivity Mouse, Rat
Antigen Map1lc3b
Immunogen A synthetic peptide from mouse microtubule-associated proteins 1A/1B light chain 3B
Quantity 150μL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Host Species Rabbit

Map1lc3b Antibody, OSM00025W, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide from mouse microtubule-associated proteins 1A/1B light chain 3B
Formulation: 150μL unpurified antibody in whole serum and no preservative
Synonyms: MAP1A/MAP1B LC3 B, Map1lc3b
Uniprot Accession: Q62625, Q9CQV6
Entrez GeneID: 64862, 67443
Gene Symbol: MAP1LC3B
Reconstitute with 150 ul of distilled water.