Please login to your online account to display your discounted pricing

anti-MAP1B (p Thr1265), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142827SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142827SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.12mg/mL
Conjugate Unlabeled
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Phosphorylated Thr1265 on MAP1B (Thr1261 in rat). Human, mouse, rat, chicken, Xenopus, fish and reptile.
Quantity 0.025mL
Gene ID (Entrez) 4131
Cross Reactivity Chicken
Cross Reactivity Fish
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Reptile
Cross Reactivity Xenopus
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Host Species Rabbit
Antigen MAP1B (p Thr1265)
Immunogen Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot No. P15205]
Format Affinity Purified
Regulatory Status RUO