Please login to your online account to display your discounted pricing

anti-MAP1B (p Thr1265), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP142827

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP142827

  • / Each
 

Description & Specifications

Specifications

Antigen MAP1B (p Thr1265)
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Concentration 0.12mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Fish
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Reptile
Cross Reactivity Xenopus
Format Affinity Purified
Formulation PBS
Host Species Rabbit
Immunogen Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot No. P15205]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4131
Test Specificity Phosphorylated Thr1265 on MAP1B (Thr1261 in rat). Human, mouse, rat, chicken, Xenopus, fish and reptile.
Target Species Chicken
Target Species Fish
Target Species Human
Target Species Murine
Target Species Rat
Target Species Reptile
Target Species Xenopus