Please login to your online account to display your discounted pricing

anti-MAP1B (p Thr1265), Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot No. P15205]
Antigen MAP1B (p Thr1265)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP142827

 
novus biologicals canada
NBP142827
Unlabeled 0.1mL Each for $329.00

NBP142827SS

 
novus biologicals canada
NBP142827SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-Prot No. P15205]
Antigen MAP1B (p Thr1265)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Test Specificity Phosphorylated Thr1265 on MAP1B (Thr1261 in rat). Human, mouse, rat, chicken, Xenopus, fish and reptile.
Gene ID (Entrez) 4131
Formulation PBS
Concentration 0.12mg/mL