Please login to your online account to display your discounted pricing

anti-MALT1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 43-57 ELLDQAPEGRGWRRL of human MALT1 (GenBank No. NP_776216.1) was used as immunogen. The immunogen sequence is 100% conserved between Malt1 isoforms a and b.
Antigen MALT1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224732

 
novus biologicals canada
NBP224732
Unlabeled 0.1mg Each for $255.80

NBP224732SS

 
novus biologicals canada
NBP224732SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 43-57 ELLDQAPEGRGWRRL of human MALT1 (GenBank No. NP_776216.1) was used as immunogen. The immunogen sequence is 100% conserved between Malt1 isoforms a and b.
Antigen MALT1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The antibody was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight 46 kDa.
Gene Accession No. Q9UDY8
Gene ID (Entrez) 10892
Formulation 100μg of antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL