Please login to your online account to display your discounted pricing

anti-MALT1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human MALT1/Paracaspase protein was used as immunogen.
Antigen MALT1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056158T

 
novus biologicals canada
NB10056158SS
Unlabeled 0.025mL Each for $144.63

NB10056158

 
novus biologicals canada
NB10056158
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human MALT1/Paracaspase protein was used as immunogen.
Antigen MALT1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity The antibody recognizes both human MALT1 isoforms a and b. Tranlocations between the MALT1 gene and other genes, can result in MALT1 chimeric (fusion) proteins of various sizes.
Gene ID (Entrez) 10892
Formulation PBS