Please login to your online account to display your discounted pricing

anti-MALT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224732

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224732

  • / Each
 

Description & Specifications

Specifications

Antigen MALT1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg of antibody in 200μL PBS containing 0.2% gelatin
Gene Accession No. Q9UDY8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 43-57 ELLDQAPEGRGWRRL of human MALT1 (GenBank No. NP_776216.1) was used as immunogen. The immunogen sequence is 100% conserved between Malt1 isoforms a and b.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10892
Test Specificity The antibody was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight 46 kDa.
Target Species Human