Please login to your online account to display your discounted pricing

anti-MALT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056158SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056158T

  • / Each
 

Description & Specifications

Specifications

Antigen MALT1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen Full-length recombinant human MALT1/Paracaspase protein was used as immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10892
Test Specificity The antibody recognizes both human MALT1 isoforms a and b. Tranlocations between the MALT1 gene and other genes, can result in MALT1 chimeric (fusion) proteins of various sizes.
Target Species Human